PEG5-carboxylic acid tert-butyl ester ≥94.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:518044-32-1
:
Formula:C₁₅H₃₀O₇
MDL Number:MFCD11041117
:322.40 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 94.0 %
Specification Test Results
Frequently Bought Together