
SB-525334 ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:356559-20-1
:
Formula:C₂₁H₂₁N₅
MDL Number:MFCD11045307
:343.43 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:156.0 to 160.0 deg-C
- Elemental analysis (Nitrogen):19.40 to 21.40 %
Specification Test Results
Frequently Bought Together