Order Entry
Portugal
ContactUsLinkComponent
 
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate ≥97.0% (by GC)
 :  TCI
 
undefined
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %
  • Acid Value:
    max. 3.0

 

Frequently Bought Together