Boc-L-Pro-OMe ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:59936-29-7
:
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD01075092
:229.3 g/mol
Boiling Pt:135°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Optical purity (LC):min. 98.0 ee%
Specification Test Results
Frequently Bought Together