4-Bromo-9,9'-spirobi[9H-fluorene] ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1161009-88-6
:
Formula:C₂₅H₁₅Br
MDL Number:MFCD16660920
:395.30 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:157.0 to 161.0 deg-C
Specification Test Results
Frequently Bought Together