Order Entry
Portugal
ContactUsLinkComponent
 
 
 
2-Bromo-1-(o-tolyl)ethanone ≥96.0% (by GC, titration analysis)
 :  TCI
 
 
 
2-Bromo-1-(o-tolyl)ethanone ≥96.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 96.0 %
  • Purity (Argentmetric Titration):
    min. 96.0 %

 

Frequently Bought Together