
3-Bromo-2-fluoro-5-methylpyridine ≥98.0% (by GC)
: TCI
:17282-01-8
:
:C₆H₅BrFN
:MFCD03095305
:190.01 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:57.0 to 61.0 deg-C
Frequently Bought Together