(S)-(-)-Butylene oxide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: (S)-(-)-1,2-Epoxybutane, (S)-(-)-Ethylethylene Oxide, (S)-(-)-Ethyloxirane
CAS Number:30608-62-9
:
Formula:C₄H₈O
MDL Number:MFCD02683445
:72.11 g/mol
Flash Pt:–22°C
UN:3022
ADR:3,II
:GHS02
:GHS07
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:-9.0 to -11.0 deg (neat)
Specification Test Results
Frequently Bought Together