
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:203866-13-1
:
Formula:C₁₀H₁₆FNO₄
MDL Number:MFCD04973957
:233.24 g/mol
Melting Pt:162°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 97.0 %
- Specific rotation [a]20/D:-55.0 to -59.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together