Order Entry
Portugal
ContactUsLinkComponent
Alloc-Val-Ala-PAB-OH ≥97.0% (by HPLC, total nitrogen)
Alloc-Val-Ala-PAB-OH ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAA3347-250MG
Supplier:  TCI
CAS Number:  
Alloc-Val-Ala-PAB-OH ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAA3347-250MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    225.0 to 229.0 deg-C
  • Specific rotation [a]20/D:
    -14.0 to -18.0 deg (C=1, DMF)

Specification Test Results

About this item

 :
 :377.44 g/mol
Storage Temperature:Freezer
CAS Number:1343407-91-9
Formula:C₁₉H₂₇N₃O₅