Order Entry
Portugal
ContactUsLinkComponent
 
AM-251 ≥97.0% (by HPLC)
 :  TCI
 
undefined
AM-251 ≥97.0% (by HPLC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Elemental analysis (Nitrogen):
    9.60 to 10.60 %

 

Frequently Bought Together