5-Acetonyl-2-methoxybenzene sulphonamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
Synonyms: 5-Acetonyl-2-methoxybenzenesulphonamide, 2-Methoxy-5-(2-oxopropyl)benzenesulphonamide
CAS Number:116091-63-5
:
Formula:C₁₀H₁₃NO₄S
MDL Number:MFCD07782136
:243.2795 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together