O-(2-Azido-4,6-O-benzylidene-2-deoxy-ɑ-D-galactopyranosyl)-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine tert-Butyl Ester ≥92.0% (by HPLC)
Catalog # TCIAA1833-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 92.0 area%
- Specific rotation [a]20/D:+87.0 to +91.0 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:658.71 g/mol
Storage Temperature:Freezer
CAS Number:878483-02-4
Formula:C₃₅H₃₈N₄O₉