
DL-Adrenaline hydrochloride ≥98.0% (by HPLC, titration analysis)
: TCI
:329-63-5
:
:C₉H₁₃NO₃
:MFCD00050562
:219.67 g/mol
:14,03619
:206-346-0
:2811
:6.1,III
:GHS06
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Water:almost transparency
Frequently Bought Together