2,3-Butanedione monoxime 98%
:57-71-6
:
:C₄H₇NO₂
:MFCD00002116
:101.11 g/mol
:185…186 °C (760 mmHg)
:74…76 °C
:Ambient
:200-348-5
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:74 to 77 °C
- Sulfated ash:≤0.1 %
- Solubility:(5 % in water) Clear to slightly hazy
- GC:≥97.5 %
Frequently Bought Together