


Ethyl-(S)-(-)-lactate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:687-47-8
:MFCD00065359
Formula:C₅H₁₀O₃
MDL Number:MFCD00004518
:118.13 g/mol
Merck Index:12,03850
Boiling Pt:154 °C (1013 hPa)
Melting Pt:–25 °C
Density:1.042 g/cm³ (20 °C)
EINECS:211-694-1
UN:1192
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:53 °C
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(25 °C) soluble
- Melting Point:-25 °C
- Molar Mass:118.13 g/mol
- Boiling Point:154 °C (1013 hPa)
- Vapor Pressure:1.6 hPa (20 °C)
- Flash Point:53 °C
- Refractive Index:1.411 (25 °C, 589 nm)
- Explosion Limit:1.5 - 16.4 %(V)
- Density:1.03 g/cm3 (20 °C)
- pH:4 (50 g/l, H2O, 20 °C)
- Ignition Point:400 °C
- Viscosity kinematic:2.7 mm2/s (25 °C)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together