Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium chloride
Sodium chloride 5 mol/l (5 N) in aqueous solution for molecular biology
Sodium chloride 5 mol/l (5 N) in aqueous solution for molecular biology
Sodium chloride 5 mol/l (5 N) in aqueous solution for molecular biology
PROMV42211000
:
Promega
:
Sodium chloride 5 mol/l (5 N) in aqueous solution for molecular biology
PROMV42211000
:
Promega
:
Pack type:
Glass bottle
Pk:
1.000 mL
Seq:
2865
:
:
58.44 g/mol
:
Ambient
:
231-598-3
:
7647-14-5
:
NaCl
:
MFCD00003477