Order Entry
Portugal
ContactUsLinkComponent
 
Anti-IgG Sheep Antibody (HRP (Horseradish Peroxidase))
  RPN4201
 :  Cytiva
undefined
Anti-IgG Sheep Antibody (HRP (Horseradish Peroxidase))
  RPN4201
 :  Cytiva

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Conjugation:
    Horseradish Peroxidase Complex, biotinylated
  • Host:
    Sheep
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Storage temperature:
    2 to 8°C
  • Pk:
    0,5 ml

 

 

For use in ELISA systems
For ELISA, it is recommended that the conjugate should be diluted in the range 1:4000 to 1:8000
Amdex conjugates are already supplied as components of the Biotrak high-sensitivity cytokine ELISAs, complete with fully-optimized protocols
Easily soluble in cold and hot water

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: