Xanthan gum, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:11138-66-2
:
MDL Number:MFCD00131256
Density:1.5 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:234-394-2
Frequently Bought Together