Order Entry
Portugal
ContactUsLinkComponent
Organic reference standard, Viridiflorol, analytical standard, Supelco®
Organic reference standard, Viridiflorol, analytical standard, Supelco®
Catalog # 72999-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Viridiflorol, analytical standard, Supelco®
Catalog # 72999-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    552-02-3
  • Description:
    Viridiflorol, ≥95,0% (GC)
  • Formula:
    C₁₅H₂₆O
  • Description:
    Viridiflorol

Specifications

About this item