Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Food and Beverage Test Systems
Sucrose assay reagent, Supelco®
Sucrose assay reagent, Supelco®
Sucrose assay reagent, Supelco®
Catalog # S1299-1VL
Supplier:
Merck
CAS Number:
Expand image
Sucrose assay reagent, Supelco®
Catalog # S1299-1VL
Supplier:
Merck
CAS Number:
Specifications
Pk:
1
Description:
Sucrose assay reagent
Specifications
About this item
Product Details
Product Details