Phytochemical standard, Isoimperatorin, analytical standard, Supelco®
68771-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:482-45-1
- Description:Isoimperatorin, ≥98,0% (HPLC)
- Formula:C₁₆H₁₄O₄
- Storage temperature:2...8 °C
- Description:Isoimperatorin