Organic reference standard, Ipriflavone, analytical standard, Supelco®
16499-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:35212-22-7
- Description:Ipriflavone, ≥98,0% (HPLC)
- Melting point:116 - 120 °C
- Formula:C₁₈H₁₆O₃
- Description:Ipriflavone