Organic reference standard, Fumagillin solution, analytical standard, Supelco®
Catalog # 37017-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:23110-15-8
- Description:Fumagillin solution
- Formula:C₂₆H₃₄O₇
- Storage temperature:−20°C
- Description:Fumagillin solution