Organic reference standard, 3-Benzylidenecamphor, analytical standard, Supelco®
Catalog # 91556-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:15087-24-8
- Description:3-Benzylidenecamphor, ≥97,0% (HPLC)
- Formula:C₁₇H₂₀O
- Description:3-Benzylidenecamphor
- Form:Neat