Organic reference standard, 2-Heptanol, analytical standard, Supelco®
Catalog # 55587-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:543-49-7
- Description:2-Heptanol, ≥98,0% (GC)
- Formula:C₇H₁₆O
- Description:2-Heptanol
- Boiling point:160...162 °C (lit.)