Order Entry
Portugal
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, (±)-Glycidol
  74595-100MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, (±)-Glycidol
  74595-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    556-52-5
  • Description:
    (±)-Glycidol
  • Environmentally Preferable:
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 + H312 - H315 - H319 - H330 - H335 - H341 - H350 - H360F
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00005147
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (±)-Oxirane-2-methanol; 2,3-Epoxy-1-propanol; Glycerolglycide
  • Molecular weight:
    74,08
  • Density:
    1,117 g/ml at 25 °C (lit.)
  • Flash point:
    72 °C
  • Boiling point:
    61 - 62 °C/15 mmHg (lit.)

 

 

  • GHS signal: GHS02, GHS06, GHS08
 : P202 - P280 - P301 + P312 - P302 + P352 + P312 - P304 + P340 + P310 - P305 + P351 + P338