Pesticide reference standard, Tralkoxydim, PESTANAL®, analytical standard, Supelco®
Catalog # 36536-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:87820-88-0
- Description:Tralkoxydim
- M-Clarity quality level:MQ100
- Formula:C₂₀H₂₇NO₃
- Description:Tralkoxydim