Order Entry
Portugal
ContactUsLinkComponent
Organic reference standard, Levofloxacin, analytical standard, Supelco®
Organic reference standard, Levofloxacin, analytical standard, Supelco®
Catalog # 40922-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Levofloxacin, analytical standard, Supelco®
Catalog # 40922-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    100986-85-4
  • Description:
    Levofloxacin
  • Formula:
    C₁₈H₂₀FN₃O₄
  • Description:
    Levofloxacin

Specifications

About this item

Packaging: Bottomless glass bottles are inserted with fused cone.