Supelco® Organic Reference Standard, Benzotriazole
Catalog # 76457-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:95-14-7
- Description:Benzotriazole
- Environmentally Preferable:
- Storage conditions:2...30 °C
- Hazard class description:H302 - H319 - H411
- Shipping conditions:STD
- MDL No.:MFCD00005699
- M-Clarity quality level:MQ100
- Synonym(s):1,2,3-Benzotriazole; 1H-Benzotriazole
- Melting point:97 - 99 °C (lit.)
- Molecular weight:119,12
- Flash point:170 °C - Closed cup
Specifications
About this item
- GHS signal: GHS07
Caution: P264 - P270 - P273 - P280 - P301 + P312 - P305 + P351 + P338