Organic reference standard, 4-Methylaminoantipyrine hydrochloride, analytical standard, Supelco®
Catalog # 92939-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:856307-27-2
- Description:4-Methylaminoantipyrine hydrochloride
- Formula:C₁₂H₁₅N₃O·HCl
- Description:4-Methylaminoantipyrine hydrochloride