Order Entry
Portugal
ContactUsLinkComponent
Screw cap, DURAN® PURE GLS 80®
Screw cap, DURAN® PURE GLS 80®
Catalog # SCOT891169109
Supplier:  DWK Life Sciences
undefined
Screw cap, DURAN® PURE GLS 80®
Catalog # SCOT891169109
Supplier:  DWK Life Sciences
Specifications
  • Ø:
    88 mm
  • Thread:
    GLS 80
  • Colour:
    White
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
The DURAN® PURE GLS 80® screw cap is made from PSU with highly inert PTFE coated silicone seal. It is designed for use with DURAN® PURE GLS 80® bottles.
  • Compatible with autoclaving and other common sterilization methods
  • With batch identification moulded into cap
  • Packaged in ISO Class 7 clean room
  • Single batch delivery
  • Certificate of Analysis and Change Control on request
Typical applications: Manufacturing of pharmaceutical products, storage of bulk drug intermediates and ingredients, transportation of APIs.