Supelco® TraceCERT® Organic Reference Standard, Potassium phosphate monobasic
Catalog # 92214-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Standards:TraceCERT®, certified reference material, 31P-qNMR Standard
- CAS No.:7778-77-0
- Description:Potassium phosphate monobasic
- Storage conditions:15...25 °C
- Shipping conditions:STD
- MDL No.:MFCD00011401
- M-Clarity quality level:MQ300
- Synonym(s):Monopotassium phosphate; Potassium dihydrogen phosphate; prim.-Potassium phosphate
- Melting point:252.6 °C (lit.)
- Formula:KH₂PO₄
- Molecular weight:136,09
- Density:2,338 g/ml at 25 °C (lit.)
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.