Calcium hydroxide 95.0-105.0% (by complexometric analysis), powder USP, BP, Ph. Eur., JP, pharma grade
:1305-62-0
:
:Ca(OH)₂
:MFCD00010901
:74.09 g/mol
:2.24 (20/4)
:215-137-3
:GHS07
- Assay (Compl.):95,0 - 100,5%
- Identity according to Pharmacopoeias:Passes Test
- Insoluble matter in HCl:≤0.5%
- Chloride (Cl):≤0.033%
- Sulphate (SO₄):≤0.4%
- Carbonate (as CaCO₃):≤5.0%
- Carbonate (USP):Passes Test
- Residual Solvents (Ph. Eur./USP):Passes Test
- Magnesium and Alkali Salts:≤4.0%
- Heavy Metals (as Pb):≤0.002%
- Elemental Impurities (ICH Q3D), Class 1, Cd:0.5 ppm
- Elemental Impurities (ICH Q3D), Class 1, Pb:0.5 ppm
- Elemental Impurities (ICH Q3D), Class 1, As:1.5 ppm
- Elemental Impurities (ICH Q3D), Class 1, Hg:1.5 ppm
- Elemental Impurities (ICH Q3D), Class 2A, Co:5 ppm
- Elemental Impurities (ICH Q3D), Class 2A, V:10 ppm
- Elemental Impurities (ICH Q3D), Class 2A, Ni:20 ppm
- Elemental Impurities (ICH Q3D), Class 2B, Au:10 ppm
- Elemental Impurities (ICH Q3D), Class 2B, Pd:10 ppm
- Elemental Impurities (ICH Q3D), Class 2B, Ir:10 ppm
- Elemental Impurities (ICH Q3D), Class 2B, Os:10 ppm
- Elemental Impurities (ICH Q3D), Class 2B, Rh:10 ppm
- Elemental Impurities (ICH Q3D), Class 2B, Ru:10 ppm
- Elemental Impurities (ICH Q3D), Class 2B, Ag:15 ppm
- Elemental Impurities (ICH Q3D), Class 2B, Pt:10 ppm
- Elemental Impurities (ICH Q3D), Class 3, Li:55 ppm
- Elemental Impurities (ICH Q3D), Class 3, Sb:120 ppm
- Elemental Impurities (ICH Q3D), Class 3, Ba:140 ppm
- Elemental Impurities (ICH Q3D), Class 3, Mo:25 ppm
- Elemental Impurities (ICH Q3D), Class 3, Cu:250 ppm
- Elemental Impurities (ICH Q3D), Class 3, Sn:600 ppm
- Elemental Impurities (ICH Q3D), Class 3, Cr:25 ppm
Frequently Bought Together