

Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) 75% (w/w) in isopropanol (IPA), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:17927-72-9
:
Formula:C₁₆H₂₈O₆Ti
MDL Number:MFCD00000031
:364.26 g/mol
Density:0.995 g/cm³ (25 °C)
Flash Pt:12 °C
UN:1219
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Specification Test Results
- Appearance (Colour):Conforms to Requirements; Yellow to Gold, Orange, Brown-Green, or Brown
- Appearance (Form):Liquid
- Infrared Spectrum:Conforms to Structure
- % Ti (After Ignition to Oxide):9.0 - 10.5%
Specification Test Results
Frequently Bought Together