Order Entry
Portugal
ContactUsLinkComponent
Zinc bromide, anhydrous ≥98%, Puriss., Fluka™
Zinc bromide, anhydrous ≥98%, Puriss., Fluka™
Certificates
Zinc bromide, anhydrous ≥98%, Puriss., Fluka™

Specification Test Results

  • Assay:
    Min. 98 %
  • Barium (Ba):
    Max. 0.001 %
  • Calcium (Ca):
    Max. 0.01 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.01 %
  • Sodium (Na):
    Max. 0.01 %
  • Lead (Pb):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

Related Information

Frequently Bought Together