Order Entry
Portugal
ContactUsLinkComponent
Hydrobromic acid ≥48%, Purum p.a.
Hydrobromic acid ≥48%, Purum p.a.
Certificates
Hydrobromic acid ≥48%, Purum p.a.

Specification Test Results

  • Assay:
    48.0 - 50.0 %
  • Refractive index (n 20/D):
    1.440 - 1.443
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Sulfate and sulfite (as SO₄):
    Max. 50 ppm
  • Vapor Pressure:
    320 psi ( 21.1 °C)

Specification Test Results

Related Information

Frequently Bought Together