Order Entry
Portugal
Orders LinkContactUsLinkComponent
T4 DNA ligase buffer pack

T4 DNA ligase buffer pack

T4 DNA ligase 10X buffer, 3×500 µl, 1,5 mL
Catalog # PROMC1263
Supplier:   Promega
T4 DNA ligase buffer pack
Catalog # PROMC1263
Supplier:  Promega
CAS Number:  
T4 DNA ligase buffer pack
T4 DNA ligase buffer pack

T4 DNA ligase 10X buffer, 3×500 µl, 1,5 mL

CAS Number:  
Price
54,60 €
Log in for availability
1 * 1,5 mL
Product Details & Documents

Buffer used with T4 DNA Ligase to join cohesive or blunt ends.

Specifications

Specifications

  • Pk:
    1,5 mL
  • Description:
    T4 DNA ligase 10X buffer
  • Pk Info:
    3×500 µl

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.