Order Entry
Portugal
ContactUsLinkComponent
Genomic DNA of E. coli O157 (EDL 933), IRMM® certified Reference Material
Genomic DNA of E. coli O157 (EDL 933), IRMM® certified Reference Material
Catalog # SIALIRMM449
Supplier:  Merck
CAS Number:  
Genomic DNA of E. coli O157 (EDL 933), IRMM® certified Reference Material
Catalog # SIALIRMM449
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Genomic DNA of E. coli O157
  • Environmentally Preferable:
Specifications
About this item
Certified for genomic DNA Eschericha coli O157, strain EDL 933 confirmed by fliC gene.