Transfection reagents, FluoMag™
About this item
FluoMag™ reagents are fluorescently-labeled magnetic nanoparticles for Magnetofection™ applications. FluoMag™ transfection reagents are as efficient as their non labelled counterparts and allow visualisation of the nanoparticles in vitro during experiments.
- Advantages of Magnetofection™ technology
- Easy cell labelling and tracking
- Non toxic
- Serum compatible
Store at +4 °C and protect from the light.
Ordering information: This reagent needs to be used with a magnetic plate, available separately.
Delivery information: Shipped at room temperature.
5 Options Available Below
Product Details & Documents
FluoMag™ reagents are fluorescently-labeled magnetic nanoparticles for Magnetofection™ applications. FluoMag™ transfection reagents are as efficient as their non labelled counterparts and allow visualisation of the nanoparticles in vitro during experiments.
- Advantages of Magnetofection™ technology
- Easy cell labelling and tracking
- Non toxic
- Serum compatible
Store at +4 °C and protect from the light.
Ordering information: This reagent needs to be used with a magnetic plate, available separately.
Delivery information: Shipped at room temperature.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- OZBIFC10100
- OZBIFN10100
- OZBIFP10100
- OZBIFS10100
- OZBIFV10100
- Cell specific:
- Neurons
- siRNA:
- Yes
- Transduction enhancer:
- Yes
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- OZBIFC10100FluoMag™-C transfection reagent corresponding to CombiMag (transfection reagent enhancer)FluoMag™-C transfection reagent corresponding to CombiMag (transfection reagent enhancer)100 µlLog in for availability1 * 100 µl97,07 €Specifications:
More
Specifications
Cat. No.OZBIFC10100Cell specificsiRNATransduction enhancer - OZBIFN10100FluoMag™-N transfection reagent corresponding to NeuroMag (neuron transfection)FluoMag™-N transfection reagent corresponding to NeuroMag (neuron transfection)100 µlLog in for availability1 * 100 µl134,83 €Specifications:
More
Specifications
Cat. No.OZBIFN10100Cell specificNeuronssiRNATransduction enhancer - OZBIFP10100FluoMag™-P transfection reagent corresponding to PolyMag (DNA transfection reagent)FluoMag™-P transfection reagent corresponding to PolyMag (DNA transfection reagent)100 µlLog in for availability1 * 100 µl185,25 €Specifications:
More
Specifications
Cat. No.OZBIFP10100Cell specificsiRNATransduction enhancer - OZBIFS10100FluoMag™-S transfection reagent corresponding to SilenceMag (siRNA delivery)FluoMag™-S transfection reagent corresponding to SilenceMag (siRNA delivery)100 µlLog in for availability1 * 100 µl87,36 €Specifications:
More
Specifications
Cat. No.OZBIFS10100Cell specificsiRNAYesTransduction enhancer - OZBIFV10100FluoMag™-V transfection reagent corresponding to ViroMag (viral transduction)FluoMag™-V transfection reagent corresponding to ViroMag (viral transduction)100 µlLog in for availability1 * 100 µl134,83 €Specifications:
More
Specifications
Cat. No.OZBIFV10100Cell specificsiRNATransduction enhancerYes
Accessories
Product Information
- Catalog #AvailabilityPrice
- Super magnetic plate, 8×12 cm
Product Information
- Catalog #AvailabilityPrice
- Mega magnetic plate, 20×26 cm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



