Order Entry
Portugal
ContactUsLinkComponent
Professional ventilated refrigerators, GKPv
Professional ventilated refrigerators, GKPv
Supplier:  Liebherr
Certificates
undefined
Professional ventilated refrigerators, GKPv
Supplier:  Liebherr
Specifications
  • Catalog No:
  • 472-1060
  • 471-0910
  • 471-1066
  • 472-1064
  • Model:
  • GKPv 1470
  • GKPv 6590
  • GKPv 6573
  • GKPv 6570
  • Cat. No.:
  • 472-1060
  • 471-0910
  • 471-1066
  • 472-1064
  • Temperature:
  • +1...+15 °C
  • −2...+16 °C
  • 1...15 °C
  • +1...+15 °C
  • Display:
  • Digital
  • Digital
  • Digital
  • Digital
  • Defrost:
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Alarm:
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Shelves/drawers:
  • 2×4 plastic-coated shelves
  • 4 plastic-coated shelves
  • 4 plastic-coated shelves
  • 4 plastic-coated shelves
  • Door type:
  • 2 solid swing doors
  • 1 solid swing door
  • 1 glazed swing door, with stainless steel frame
  • 1 solid swing door
  • Door lock:
  • Yes
  • Yes
  • Yes
  • Yes
  • Specifications:
  • Electronic temperature controller
  • Electronic temperature controller, multi-function digital display and RS485 interface
  • Electronic temperature controller
  • Electronic temperature controller
  • W×D×H ext.:
  • 1430×830×2150 mm
  • 700×830×2150 mm
  • 700×830×2150 mm
  • 700×830×2150 mm
  • Weight:
  • 239 kg
  • 180 kg
  • 144 kg
  • 180 kg
Specifications
Related Information
Product Information
Frequently Bought Together