Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Catalog # VWRS20821.410
Supplier: VWR Collection
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.8 %
- Assay (V/V):Min. 99.85 %
- IR Spectrum:Passes test
- Solubility in water:Passes test ACS
- Volatile impurities:Passes test Ph.Eur.
- Acidity:Max. 0.0001 meq/g
- Alkalinity:Max. 0.0001 meq/g
- Boiling point:78.0 - 78.5 °C
- Colouration:Max. 10 APHA
- Density (20/20):0.790 - 0.793
- n 20/D:1.360 - 1.362
- Substances coloured by H2SO4:Max. 20 APHA
- Aldehydes (as CH3CHO):Max. 5 ppm
- Aldehydes + ketones (as CH3CHO):Max. 10 ppm
- iso-Amyl alcohol:Max. 0.05 %
- Benzene:Max. 2 ppm
- Evaporation residue:Max. 5 ppm
- Formaldehyde:Max. 1 ppm
- Furaldehyde:Max. 1 ppm
- Higher alcohols (standard mixture):Max. 0.02 %
- Ketones (as CH3COCH3):Max. 10 ppm
- Methanol:Max. 100 ppm
- 2-Propanol:Max. 30 ppm
- Substances reducing KMnO4 (as O):Max. 3 ppm
- Total heavy alcohols:Max. 100 ppm
- Water:Max. 0.1 %
- Al (Aluminium):Max. 0.02 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.02 ppm
- Ca (Calcium):Max. 0.2 ppm
- Cd (Cadmium):Max. 0.02 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.01 ppm
- Cu (Copper):Max. 0.1 ppm
- Fe (Iron):Max. 0.05 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0.01 ppm
- Na (Sodium):Max. 0.5 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.05 ppm
- Sn (Tin):Max. 0.05 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.1 ppm
- Absorbance (240 nm) (5 cm):Max. 0.4
- Absorbance (250-260 nm) (5 cm):Max. 0.3
- Absorbance (270-340 nm) (5 cm):Max. 0.1
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
About this item
:
:46.07 g/mol
Merck Index:12,03806
Boiling Pt:78.3 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.7895 g/cm³ (20 °C)
Flash Pt:12 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
REACH:01-2119457610-43
:GHS02
:GHS07
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568