VWR® PerfectBlue Maxi M, Gel system
700-0880
- Description:PerfectBlue gel system Maxi M
- Model:Maxi M
- Gel volume:250 ml
- Buffer volume:~2300 ml
- Run time:≥60 h
- Gel size W×L:200×250 mm
- Sample capacity:8 – 168
- Unit W×L×H:290×370×100 mm
- Pk:1
Wider than the Maxi S Plus, the Maxi M model can accommodate up to 168 samples but still with an economical footprint.
*Based on an agarose gel of approximately 5 mm thickness.
: Supplied with 3 combs (1,5 mm thickness, 16, 24, 36 teeth), lid, gel tray, 3-point levelling platform, end gates, tank and cables.