


Cadmium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10108-64-2
:
Formula:CdCl₂
MDL Number:MFCD00010916
:183.32 g/mol
Merck Index:12,01653
Boiling Pt:967 °C (1013 hPa)
Melting Pt:565 °C
Density:4.05 g/cm³ (20 °C)
UN:2570
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:233-296-7
:GHS06
:GHS08
:GHS09
Frequently Bought Together