Zinc bromide, anhydrous 98%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7699-45-8
:
Formula:Br₂Zn
MDL Number:MFCD00011294
Merck Index:13,10182
Boiling Pt:697 °C (760 mmHg)
Melting Pt:394 °C
Density:4.22 g/cm³
Storage Temperature:Ambient
EINECS:231-718-4
ADR:8,II
:225.2 g/mol
UN:3260
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:White to off-white Powder and/or chunks
- Titration Argentometric:≥97.5 %
- Water:≤2 % (K.F.)
- Sulfate (SO4):≤100 ppm
- Barium (Ba):≤10 ppm
- Calcium (Ca):≤100 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤50 ppm
- Magnesium (Mg):≤100 ppm
- Potassium (K):≤100 ppm
- Sodium (Na):≤100 ppm
Specification Test Results
Frequently Bought Together