Entrada de Pedido
Portugal
Orders LinkContactUsLinkComponent
Sulfato de prata, Supelco®

Sulfato de prata, Supelco®

Corrosive
Environment
Supplier:   Merck
Sulfato de prata, Supelco®
Supplier:  Merck
Certificates
Sulfato de prata, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  Disilver sulphate
  • CAS Number:10294-26-5
  • Formula:Ag₂SO₄
  • MDL Number:MFCD00003407
  • Molecular Weight:311.8 g/mol
  • Merck Index:13,08603
  • Boiling Pt:1085 °C (1013 hPa)
  • Melting Pt:655 °C
  • Density:5,45 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
  • EINECS:233-653-7
  • ADR:9,III
  •  :Nível de qualidade M-Clarity™ = MQ100
  • UN:3077

Documents

Specifications for Product Family

Specification Test Results

  • Solubility:
    8 g/l (25 °C)
  • Melting Point:
    655 °C
  • Molar Mass:
    311.8 g/mol
  • Bulk Density:
    1200 kg/m3
  • Density:
    5.45 g/cm3 (20 °C)
  • pH:
    5 - 6 (5 g/l, H2O, 25 °C)
Product Family Options

Product Information

  • Tipo de embalagem
    Und.
    Availability
    Price
  • Frasco de plástico para sólidos
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 329,00 €
    329,00 €
    1 * 50 g
       329,00 €

  • Frasco de plástico para sólidos
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 1 410,00 €
    1 410,00 €
    1 * 250 g
       1 410,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.