Copper(I) bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7787-70-4
:
Formula:BrCu
MDL Number:MFCD00010969
:143.45 g/mol
Merck Index:14,2659
Boiling Pt:1345 °C
Melting Pt:492 °C
Density:4.98
Storage Temperature:Ambient
EINECS:232-131-6
ADR:9,III
:GHS07
Related Information
Frequently Bought Together