
Ninhydrin, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:485-47-2
:
Formula:C₉H₆O₄
MDL Number:MFCD00003791
:178.14 g/mol
Merck Index:13,06582
Boiling Pt:351 °C (1013 hPa)
Melting Pt:250…258 °C
Density:0.862 g/cm³ (20 °C)
EINECS:207-618-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:min 99%
- Identification and melting point:Passes test
- IR Spectrum:Passes test
- Solubilty in water (10 g/l):Passes test
- Suited for amino acid reagent:Passes test
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC25905.107 spec_VWRC25905.107 spec_VWRC25905.153 spec_VWRC25905.153 spec_VWRC25905.153
Product specification
More
VWR® Chemicals ACS or Reag. Ph. Eur.
Product Literature
Frequently Bought Together