Order Entry
Portugal
ContactUsLinkComponent
Ammonium iodide ≥99.0% (by argentometric titration), Puriss. p.a. ACS, Fluka™
Ammonium iodide ≥99.0% (by argentometric titration), Puriss. p.a. ACS, Fluka™
Certificates
Ammonium iodide ≥99.0% (by argentometric titration), Puriss. p.a. ACS, Fluka™

Specification Test Results

  • Assay:
    99.0 - 100.0 %
  • pH (5 %, 20°C):
    4.5 - 6.5
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying
  • Residue on ignition (SO₄):
    Max. 0.02 %
  • Water insoluble matter:
    Max. 0.005 %
  • Barium (Ba):
    Max. 20 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 1 ppm
  • Iron (Fe):
    Max. 2 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride and bromide (as Cl):
    Max. 50 ppm
  • Total S (as SO₄):
    Max. 20 ppm
  • Phosphate (PO₄):
    Max. 10 ppm

Specification Test Results

Related Information

Frequently Bought Together