Ammonium iodide ≥99%, Puriss., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:12027-06-4
:
Formula:NH₄I
MDL Number:MFCD00011424
Merck Index:13,00531
Melting Pt:551 °C
Density:2.510 g/cm³ (20 °C)
:144.94 g/mol
EINECS:234-717-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99.0 %
- Loss on drying (105°C):Max. 1 %
- Residue on ignition (as sulfate):Max. 0.1 %
- pH (5 %, 20°C):4.5 - 6.5
- Arsenic (As):Max. 0.0005 %
- Iron (Fe):Max. 0.001 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride and bromide (as Cl):Max. 0.02 %
- Iodate (IO₃):Max. 0.01 %
- Sulfate (SO₄):Max. 0.01 %
- Thiosulfate (S₂O₃):Max. 0.01 %
Specification Test Results
Frequently Bought Together